Imprimer
Entonnoirs de filtration
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07200325 07200425
LENZ07200325EA 45.4 EUR
LENZ07200325 LENZ07200425
Entonnoirs de filtration
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Funnels, made of boroslicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.