Imprimer
Colonnes HPLC d'échange anionique, PRP-X110 et X110S
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
79775 79776 79732 79738 79743 79731 79735 79741 79726 79727 79728 79729 79734 79733 79293
554-0453EA 973 EUR
554-0453 554-0454 554-0411 554-0417 554-0422 554-0410 554-0414 554-1826 546-8102 554-0407 554-0408 554-0406 554-0413 554-0412 HAMI79293
Colonnes HPLC d'échange anionique, PRP-X110 et X110S
Colonnes pour chromatographie
PRP-X110 columns are designed for use in systems employing suppressed/non-suppressed conductivity, electrochemical, UV, and ICP-MS detection.
  • Greatly reduce sample pretreatment
  • Improves the accuracy and precision of results
  • No calculations, just mix and run samples