Imprimer
Conteneurs, col large, avec bouchon à charnière, Corning® Gosselin™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FL300-02 FL60-05 FL100-04 FL300-04 FL100-05 FL60-04
215-3269EA 490 EUR
215-3269 215-3270 215-1285 GOSSFL300-04 GOSSFL100-05 GOSSFL60-04
Conteneurs, col large, avec bouchon à charnière, Corning® Gosselin™
Récipients de collecte d'échantillon Conteneurs à usage universel
Made of PP, natural or coloured.
  • Available in sterile or under aseptic conditions packed versions
  • Very good chemical and temperature resistance
  • Graduated
Ideal for sampling, storage and transport of liquid and solid samples.
Sized to provide adequate volume for milk sampling. It is shock resistant and is assured sterile 10-3.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904