Imprimer
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
55019-50 55019-52 55019-54 55019-56
HAMI55019-50EA 956 EUR
HAMI55019-50 HAMI55019-52 HAMI55019-54 HAMI55019-56
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Pipettes Pipetteurs
Hamilton SoftGrip pipettes are available as 8-channel and 12-channel adjustable volume manual pipettes in two volume ranges covering 5 to 50 µl and 30 to 300 µl.
  • Long tip ejector lever and movable lower body can be rotated a full 360º relative to the handle to provide maximum comfort for left- or right-handed use
  • Colour-coded plunger buttons and labels for quick volume identification
  • Easily activated tip ejector lever releases all the tips in single stroke
  • Can be autoclaved without the need for disassembly or recalibration
Please note: Accuracy and precision is guaranteed when used with Hamilton Universal AdvanTip™ precision pipette tips.