Imprimer
AdvanTip™ Pipette Tips
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11003-15 68009-20 68009-21 68009-22 68009-11
HAMI11003-15EA 111 EUR
HAMI11003-15 HAMI68009-20 HAMI68009-21 HAMI68009-22 HAMI68009-11
AdvanTip™ Pipette Tips
Pointes de pipettes
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
  • Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
  • Certified free from RNase, DNase, and pyrogens
  • Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
  • Low retention tips (only some tips): Feature walls that maximise
En savoir plus