Imprimer
Reaction vessels, round bottom, with flat flange
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06008288 06008294 06008488 06008494 06008588 06008594 06008149-V2 06008158-V2 06008170-V2 06008273-V2 06008279-V2 06008285-V2 06008485-V2 06008585-V2
LENZ06008288EA 2000 EUR
LENZ06008288 LENZ06008294 LENZ06008488 LENZ06008494 LENZ06008588 LENZ06008594 LENZ06008149-V2 LENZ06008158-V2 LENZ06008170-V2 LENZ06008273-V2 LENZ06008279-V2 LENZ06008285-V2 LENZ06008485-V2 LENZ06008585-V2
Reaction vessels, round bottom, with flat flange
Récipients de réaction Flacons à réactions
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat flange with groove and drain valve.