Imprimer
Dropping bottles, series 302
Fournisseur: Kautex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
302-81015 302-92676 302-99529
KAUT302-81015EA 88.1 EUR
KAUT302-81015 KAUT302-92676 KAUT302-99529
Dropping bottles, series 302
Flacons Flacons compte-gouttes
HDPE. Round. Natural colour.
  • Dropping tip closure
  • Separate screw closure
  • Resealable
  • Not suitable for food
  • Stable and handy shape
  • Halal certified
Suitable for adhesives and many other products.