Imprimer
Pycnometer, according to Gay-Lussac, unadjusted
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05630010 05630013 05630022 05630028 05630037
LENZ05630010EA 36.1 EUR
LENZ05630010 LENZ05630013 LENZ05630022 LENZ05630028 LENZ05630037
Pycnometer, according to Gay-Lussac, unadjusted
Specific Gravity Bottles
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • Stopper NS 10/19 with capillary
  • Top of stopper is grounded and polished
  • Nominal capacity printed on the bottom
According to DIN 12797 and ISO 3507