Imprimer
Bottles, narrow neck, square, HDPE, with thread, 'SafeGrip', series 310
Fournisseur: Kautex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
310-85405 310-85681
215-0099EA 12.1 EUR
215-0099 KAUT310-85681
Bottles, narrow neck, square, HDPE, with thread, 'SafeGrip', series 310
Flacons Flacons en plastique
HDPE. Natural colour or black.
  • UN approval
  • For hazardous liquids
  • GL45 neck
  • Leakproof
  • Halal certified
UN approved for liquids. Optimized handle with increased size for safe and comfortable handling.
UN approval: UN 3H1/X1.9/250