Imprimer
Water estimators, Dean-Stark
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05600002 05600000 05600004
661-1107EA 286 EUR
661-1107 LENZ05600000 LENZ05600004
Water estimators, Dean-Stark
Distillation Components Distillation Sets
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Water estimators acc. to DIN 12420 (xylol method), capacity: 10:0,1 ml, with round bottom flask 500 ml, ground joint connections: NS29/32, measuring tube with or without stopcock, with Liebig condenser, jacket length: 400 mm.