Imprimer
VWR® Vacuum Desiccators
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
467-0088EA 548 EUR
467-0088 467-0086 467-0087 467-0089 467-0090
VWR® Vacuum Desiccators
Desiccators Desiccators, Glass
Borosilicate 3.3 glass.
  • Precisely ground flat flanges and interchangeable tubulated lids
  • Standard ground joint NS 24/29 junction tube in the lid, complete with stopcock plus PTFE spindle
  • Base and lid with flat flange ground to fit
  • High mechanical, thermal and chemical resistance
Due to the increased wall thickness, these desiccators can be used up to the maximum vacuum that is technically possible.
Certifications: DIN 12491