Imprimer
Multidishes, Nunc™ UpCell™ surface
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174897 174898 174899 174900 174901
734-2377EA 359 EUR
734-2377 734-2378 734-2379 734-2380 734-2252
Multidishes, Nunc™ UpCell™ surface
Microplaques Microplaques pour culture cellulaire
The UpCell™ surface is a temperature-responsive cell culture surface that enables harvesting of cells with high viability and intact surface proteins for culture passaging, single-cell analyses and cell transplantation research. The UpCell™ surface enables harvesting of cell sheets and creation of 3D tissue models held together by normal cell junctions and extracellular matrix deposited by the cells.
  • No trypsinisation - cell surface proteins are preserved
  • High cell viability without scraping
  • Releases adherent cells by reduction of temperature of the cell culture
  • Requires minimal hands-on time