Imprimer
Adapters, receiver, vacuum, straight or bent, with GL thread
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05275014 05272011 05272012 05272013 05272014 05275011 05275012 05275013
LENZ05275014EA 65 EUR
LENZ05275014 LENZ05272011 LENZ05272012 LENZ05272013 LENZ05272014 LENZ05275011 LENZ05275012 LENZ05275013
Adapters, receiver, vacuum, straight or bent, with GL thread
Distillation Components Distillation Adapters
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receiver adapters, straight, with NS socket and NS cone, with drain tip, vacuum connection: thread GL 18.
Certifications: DIN 12594