Imprimer
Measuring cylinders, tall form, class A, PMP
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7520170 7520175 7520180 7520186 7520190 7520193 7520194
HIRS7520170EA 33.6 EUR
HIRS7520170 HIRS7520175 HIRS7520180 HIRS7520186 HIRS7520190 HIRS7520193 HIRS7520194
Measuring cylinders, tall form, class A, PMP
Éprouvettes Éprouvettes graduées
PMP, transparent, tall form, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • With embossed scale
  • Autoclavable - Does not affect accuracy
Certifications: DIN 12 681 / ISO 6706