Imprimer
Condensers, high efficient, four coil
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05255000 05255050
LENZ05255000EA 1810 EUR
LENZ05255000 LENZ05255050
Condensers, high efficient, four coil
Condenseurs
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
High-efficient condensers with four cooling coils, cooling surface: 2,350 cm².