Imprimer
Burettes, lateral stopcock, Schellbach
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3210160 3210170 3210175 3210360 3210370 3210375
612-9821EA 98.4 EUR
612-9821 612-9822 612-9823 HIRS3210360 HIRS3210370 HIRS3210375
Burettes, lateral stopcock, Schellbach
Burettes
Burette, class B, DURAN® borosilicate glass 3.3, with blue graduation.
  • Lateral glass stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
Certifications: Calibrated for EX, DIN EN ISO 385.