Imprimer
Multiple adapters
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05015011 05015024 05015044 05015023 05015033
LENZ05015011EA 80.7 EUR
LENZ05015011 LENZ05015024 201-2221 LENZ05015023 LENZ05015033
Multiple adapters
Distillation Components Multiple Distillation Adapters
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).