Imprimer
Disposable earplugs, E-A-RSoft™ FX
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ES01020 ES01021
111-0835EA 104 EUR
111-0835 MMMAES01021
Disposable earplugs, E-A-RSoft™ FX
Earplugs Disposable Earplugs
This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industry. Custom polyurethane foam formulation provides slow recovery and extreme softness for optimum comfort.
  • Flange shape serves as an effective insertion device when rolled down
  • Easy to fit into the ear canal
  • Available with or without cord
  • Compatible with other PPE
SNR: 39 dB