Imprimer
Compact automatic burettes, SILBERBRAND
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
23829 23830
BRND23829EA 173 EUR
BRND23829 BRND23830
Compact automatic burettes, SILBERBRAND
Burettes
Borosilicate glass 5.4, amber burette, amber PE bottle, class B.
  • Modular concept - all components are replaceable
  • Calibrated to deliver 'TD, Ex'
  • White graduations and automatic zeroing
25 ml: Error limit according to class B, DIN EN ISO 385.
50 ml: DIN EN ISO 385, class B.
Informations de commande: Please order supports, burette clamps and rubber bellows separately.
Informations de livraison: Burette supplied with 1000 ml PE reservoir bottle with base and PVC filling tube.