Imprimer
Compact automatic burettes, Schellbach, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
23819 23820 23821
612-7170EA 151 EUR
612-7170 612-7171 612-7172
Compact automatic burettes, Schellbach, BLAUBRAND®
Burettes
Borosilicate glass 3.3 bottle, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe
  • Detachable stopcock
Certifications: DIN EN ISO 385
Informations de commande: Please order burette clamps, supports and rubber bellows separately.
Informations de livraison: Supplied as burette tube, stopcock with precision tip, filling tube (PVC) and 1000 ml PE bottle