Imprimer
Sleeves for conical ground joints
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
01101514 01101519 01101524 01101529 01101545
LENZ01101514EA 36.5 EUR
LENZ01101514 LENZ01101519 LENZ01101524 LENZ01101529 LENZ01101545
Sleeves for conical ground joints
Manchons et raccords pour joints NS Manchettes pour rodage de raccordement
Disposable ground joint sleeves. thin walled, made of PTFE.
  • Sleeves have a wall thickness of 0,05 mm
  • Usable to 260 °C
  • Outstanding chemical resistance