Imprimer
Filter cartridges, Sartolon®
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5102507H1 5102507H2
SARP5102507H1EA 409 EUR
SARP5102507H1 SARP5102507H2
Filter cartridges, Sartolon®
Filtres Cartouches filtrantes
Sartolon® filter cartridges are designed for broad chemical compatibility for specific applications in the pharmaceutical and chemical industry. The double-layer design leads to superior filtration performance compared to other nylon membrane filters. Ideal for filtration of a broad range of solvents and solvent-containing liquids.
  • Withstand high differential pressures in either the forward or reverse direction of flow
Membrane: Hydrophilic nylon; support layers: PP; end cap, core and cage: PP; O-rings: silicone
Maximum differential pressure: five bar at 20 °C; two bar at 80 °C