Imprimer
Surface protection papers, LabSorb
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FT-1-601400100 FT-1-601400050 FT-1-601600050 FT-2601480600K FT2601-460570K FT1-601-460050 FT1-601-600100
515-1630EA 399 EUR
515-1630 515-1629 515-1628 515-1633 SARTFT2601-460570K SARTFT1-601-460050 SARTFT1-601-600100
Surface protection papers, LabSorb
Surface Protectors Bench Protectors
The LabSorb protection papers are highly absorptive papers coated on one side with polyethylene.
  • Prevent radioactive contamination of work surfaces (in radiochemical laboratories)
  • Protect laboratory bench surfaces from spillage or splashes of liquids by preventing absorption and seepage of these liquids
  • Line animal cages for protection and improvement of hygiene
  • Protect objects from falling on hard surfaces: reduce impact
Used with the cellulose side up, the paper absorbs liquids, which are stopped by their polyethylene layer and thus prevented from soaking through. Used with the polyethylene side up, the papers are highly useful for recovering valuable or toxic liquids.
Basis weight: 140 g/m²; Absorption: 150%