Imprimer
Station de pointes de pipette, Corning® DeckWorks™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4141 4142 4143 4144
CORN4141EA 393 EUR
CORN4141 CORN4142 CORN4143 612-4196
Station de pointes de pipette, Corning® DeckWorks™
Pointes de pipettes
These tip stations maximise bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
  • Stations de pipetage multi-canaux et monocanal
  • Conception de plateau à 5 couches (2×96) adaptée au format de la plaque de micro-titrage
  • Plate-forme de pipetage stable s'adapte facilement aux environnements de hotte stérile
  • Couvercles pouvant être clipsés pour exposer un côté (96 pointes) uniquement
Les applications incluent les procédures pré et post PCR, la culture cellulaire et les dosages nécessitant de grandes quantités de pointes.
Certified Free of: RNase, DNase, Pyrogens, PCR Inhibitors.
Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of seven sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Informations de commande: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload and reload starter kit configurations.