Imprimer
Filtres sans seringue, Autovial™, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AV125EAQU AV125ENAO AV125EORG AV125UAQU AV125UGMF AV125UNAO AV125UORG AV115NPEORG AV115NPUAQU AV115NPUORG AV115UGMF AV125NPUAQU AV525UAQU AV525UORG AV525UNAO
514-0013EA 426 EUR
514-0013 516-0452 514-0014 514-0015 514-0016 516-0460 514-0017 514-0326 516-0448 514-0327 516-0451 516-0454 516-0463 516-0465 516-0464
Filtres sans seringue, Autovial™, Whatman™
Filtres Dispositifs de filtration sans seringue
The Whatman Autovial Syringeless Filter replaces multiple syringe-coupled sample filtration devices with a single, disposable unit.
  • Available in 5 or 12 ml capacity
  • Single unit convenience – pre-assembled and easy to load
  • Self-contained, preventing filter pop-off with hazardous samples
  • Range of sterile or nonsterile membrane media available
  • Built-in air purge maximizes sample recovery
  • Glass fiber or polypropylene prefilter in selected 12 ml models – for difficult-to-filter samples
  • Choice of glass fiber or polypropylene prefilters
Autovial from Cytiva's business combines Whatman quality membranes and devices with an all-in-one syringeless filter design.
Filtration involving multiple consumables like syringe filters, vials, and septa increases storage space required for these components and time required to perform sample filtration. Cytiva's Autovial combines all of these components into a single consumable.
Autovial devices are comprised of two parts: a graduated filter barrel and a plunger. Sample is poured into the filter barrel, and the plunger is inserted until the bottom is securely in place. The tip of the device is placed into the mouth of an autosampler vial and the plunger compressed.
* Sterile