Imprimer
Cryotubes, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5001-0050 5001-0020 5001-0012 5001-1020
479-2502EA 168 EUR
479-2502 479-2501 479-2500 479-2504
Cryotubes, Nalgene®
Fioles
Adaptés au stockage réfrigéré standard (+4 °C) jusqu'à l'azote liquide en phase gazeuse.
  • Black barcode on white background for high contrast and readability even in high-frost conditions
  • Human readable code below the barcode for easy visual identification without a barcode reader
  • External thread, conical bottom, self-standing
  • Non cytotoxic, DNase- and RNase-free
En PPCO, stériles.
System 100™ vials with PP closure and internal silicone gasket allow for maximum storage capacity.