Acétate de phénylmercure ≥98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Purity: 98%
CAS No.: 62-38-4
Molecular Formula: C8H8HgO2
Formula Weight: 336.74
Physical Form: Powder
Density: 2.4
Appearance: White to yellow
Odour: Odourless
Melting Point: 146-153°
Solubility: Slightly soluble in water. Soluble in alcohol, benzene and glacial acetic acid
Beilstein Reference: 3662930
Merck Reference: 14,7300
UN No.: 1674
MDL No.: MFCD00008691
Formule:
C₈H₈HgO₂ Poids moléculaire: 336.74 g/mol Point de fusion: 146…153 °C Densité: 2.4 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00008691 Numéro CAS: 62-38-4 EINECS: 200-532-5 UN: 1674 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...