Dopamine chlorhydrate 99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
Chlorure de 3,4-dihydroxyphénéthylammonium
Purity: 99%
CAS No.: 62-31-7
Molecular Formula: C8H11NO2.HCl
Formula Weight: 189.64
Storage Temperature: 2°C to 8°C
Melting Point: 243-250°
Beilstein Reference: 3656720
Merck Reference: 14,3421
MDL No.: MFCD00012898
A neurotransmitter and vasopressor that modulates cortical activation
Formule:
C₈H₁₂ClNO₂ Point de fusion: 243…250 °C Densité: 1,356 g/cm³ (20 °C) |
Numéro MDL:
MFCD00012898 Numéro CAS: 62-31-7 EINECS: 200-527-8 Indice Merck: 14,3421 |