Imprimer
Pointes universelles de pipettes, Corning® DeckWorks™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4120 4121 4122 4123 4124 4117 4119 4118 4115 4116 4148 4149 4150 4151 4152 4147
613-2977EA 341 EUR
613-2977 613-2978 612-4195 613-2979 612-4197 613-2974 613-2976 613-2975 613-2972 613-2973 613-2981 613-2982 613-2983 613-2984 613-2985 613-2980
Pointes universelles de pipettes, Corning® DeckWorks™
Pointes de pipettes
En PP, graduées. Le système DeckWorks™ est conçu pour optimiser l'espace de stockage et réduire la production et les déchets de matière plastique. Les portoirs à charnières et les recharges de ce système sont fabriqués en plastique recyclé.
  • Stériles ou non stériles
  • Exemptes de DNase, RNase et d'ADN humain
  • Apyrogènes
Corning universal fit tips in hinged DeckWorks racks are available in 10, 200, 300 and 1000 µl volumes. The 10, 300 and 1000 µl tips are extended length and allow for improved access into micro (1,5 ml) and standard conical bottom (15/50 ml) centrifugre tubes. Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system.
Corning DeckWorks low binding tips are available in 10, 200 and 300 µl sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
The entire DeckWorks system is comprised of seven sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tip s are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Certifications: Corning DeckWorks pipette tips are also available in bulk, barrier hinged rack, reload, reload starter kit, and tip station configurations.