Imprimer

Bromoéthane ≥98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): Bromic ether, Hydrobromic ether

A12804.0E A12804.22 A12804.36 A12804.PG
A12804.0EEA 133 EUR
A12804.0E A12804.22 A12804.36 A12804.PG
Bromoéthane ≥98%
Bromoéthane

Purity: 98% CAS No.: 74-96-4 Molecular Formula: C2H5Br Formula Weight: 108.97 Flash Point: -23°(-9°F) Physical Form: Clear liquid Density: 1.461 Refractive Index: 1.4235 Sensitivity: Light Sensitive Appearance: Colourless Odour: Ether-like Melting Point: -119° Boiling Point: 37-39° Beilstein Reference: 1209224 Merck Reference: 14,3771 UN No.: 1891 MDL No.: MFCD00000232

Formule: BrH₂CCH₃
Poids moléculaire: 108.97 g/mol
Point d'ébullition: 37…39 °C
Point de fusion: –119 °C
Densité: 1.461
Point éclair: –23 °C (–9 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00000232
Numéro CAS: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1 II
Indice Merck: 13,03807

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR