Imprimer
Sodium molybdate dihydraté 99.5-103.0% ACS
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
12214.22 12214.36
12214.22EA 76.9 EUR
12214.22 12214.36
Sodium molybdate dihydraté 99.5-103.0% ACS
Sodium molybdate dihydraté
Purity: 99.5-103.0% CAS No.: 10102-40-6 Molecular Formula: MoNa2O4.2H2O Formula Weight: 241.95 (205.92anhy) Physical Form: Powder Density: 2.71 Appearance: White Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water Merck Reference: 14,8645 MDL No.: MFCD00149170
Formule: Na₂MoO₄·2H₂O
Poids moléculaire: 241.95 g/mol
Point de fusion: 100 °C (-2H₂O)
Densité: 2.71
Température de stockage: Température ambiante
Numéro MDL: MFCD00149170
Numéro CAS: 10102-40-6
EINECS: 231-551-7
Indice Merck: 14,8645