Imprimer
Acide sulfurique 0.05 mol/l (0.1 N) en solution aqueuse, Titripur® solution volumétrique, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M-Clarity™ (Niveau de Qualité) = MQ100
1.09074.1000 1.09074.5000 1.09074.9010 1.09074.4000 1.09074.9013
1.09074.1000EA 29.5 EUR
1.09074.1000 1.09074.5000 1.09074.9010 1.09074.4000 1.09074.9013
Acide sulfurique 0.05 mol/l (0.1 N) en solution aqueuse, Titripur® solution volumétrique, Supelco®
Acide sulfurique
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.
Formule: H₂SO₄
Poids moléculaire: 98.08 g/mol
Point d'ébullition: 330
Point de fusion: 10.38
Densité: 1.01 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064589
Numéro CAS: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III