Imprimer
Cuves de coloration avec couvercles.
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
474300 474305
BRND474300EA 170 EUR
BRND474300 BRND474305
Cuves de coloration avec couvercles.
Pots Cuves de coloration
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Informations de commande: Trough and tray (insert) have to be ordered separately.