Propanediol-1,2 99,5 ACS
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
Purity: 99.5%
CAS No.: 57-55-6
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 107°(225°F)
Physical Form: Liquid
Density: 1.036
Refractive Index: 1.4320
Sensitivity: Hygroscopic
Odour: Nearly Odourless
pH: neutral
Melting Point: -60°
Boiling Point: 186-188°
Solubility: Miscible with water, acetone, chloroform. Soluble in ether
Beilstein Reference: 1340498
Merck Reference: 14,7855
MDL No.: MFCD00064272
Common Applications: As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils.
Formule:
CH₃CH(OH)CH₂OH Poids moléculaire: 76.1 g/mol Point d'ébullition: 186…188 °C Point de fusion: –60 °C Densité: 1.036 Point éclair: 107 °C (225 °F) Température de stockage: Température ambiante |
Numéro MDL:
MFCD00064272 Numéro CAS: 57-55-6 EINECS: 200-338-0 Indice Merck: 14,7855 |