Imprimer
Flacons à centrifuger, PC/PPCO, super rapides, avec bouchon, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3140-1006 3141-1006
525-0466EA 1150 EUR
525-0466 525-0467
Flacons à centrifuger, PC/PPCO, super rapides, avec bouchon, Nalgene®
Flacons Flacons à centrifuger
Super-speed centrifuge bottles are designed to boost productivity and pellet recovery in super speed large volume gross separation applications.
  • Process a full liter in each bottle at speeds up to 15810×g
  • Smooth inner walls reduce sheering in mammalian culture separations
  • PC (for clarity and mechanical strength) and PP (for chemical resistance)
  • Recommended operating temperature range 4 to 22 °C
  • Storage temperature: –70 °C
Ideal for bacterial, yeast and tissue isolations; improved viral harvesting in vaccine production; reduced shearing in mammalian culture separations.
Certifications: Compliance with USP Class VI, non-cytotoxic.
Informations de livraison: Centrifuge bottles are supplied with 3 - piece sealing cap assembly, polypropylene plug, closure and silicone gasket.