Imprimer
Tuyaux, en polyuréthane, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8030-0060 8030-0120
NALG8030-0060EA 90.6 EUR
NALG8030-0060 224-2031
Tuyaux, en polyuréthane, Nalgene®
Tuyaux Transfer Tubing
Polyurethane, translucent.
  • Contains no plasticizers and only low levels of extractables for high-purity applications
  • Superior chemical resistance to fuels, oils, and some solvents (not recommended for aqueous solutions)
  • Resistant to atmospheric ozone, aliphatic hydrocarbons and petroleum products for longer life and versitility
  • This transparent, non-autoclavable tubing is marked every 12" for cutting and can be gas sterilized
  • Ideal for peristaltic pump applications
Polyurethane tubing, which exhibits excellent abrasion and flex-fatigue resistance, as well as low-temperature resilience.