Imprimer
Tuyaux, en silicone, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8061-3070 8061-3130 8061-3190
NALG8061-3070EA 1210 EUR
NALG8061-3070 NALG8061-3130 NALG8061-3190
Tuyaux, en silicone, Nalgene®
Tuyaux Transfer Tubing
Braided platinum-cured silicone tubing.
  • Contains no phthalate plasticisers or peroxides to leach out into transported fluid
  • Tasteless, odorless, non-cytotoxic, non-pyrogenic and non-hemolytic
  • Autoclavable; also can be sterilized by gamma radiation, ETO and chemical procedures
  • Durometer hardness: Shore A, 65
Tubing for a variety of pump and transfer applications, including pharmaceutical, laboratory, bioprocess manufacturing and food and beverage applications. This durable, high-purity tubing features an interwoven polyester braid for added safety, pressure and kink resistance.
Certifications: Meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition requirements for plastic materials; and 3A sanitary standards.