Imprimer
Measuring cylinders
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-3837EA 68.4 EUR
612-3837 612-3840 612-3838 612-3836 612-3835 612-3834 612-3833 612-3832 612-3839
Measuring cylinders
Cylinders Measuring Cylinders
Borosilicate 3.3 glass, class A, tall form.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Blue graduations with ring marks at major graduations
  • Certificate of conformity with batch number
DIN 12680
Certifications: DIN 12680
Informations de commande: Batch number and batch certificate included.