Imprimer
VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-3826EA 79.7 EUR
612-3826 612-3825 612-3824 612-3823 612-3822 612-3821 612-3828 612-3829 612-3827 612-3831 612-3830
VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Flasks Volumetric Flasks
Borosilicate 3.3 glass, amber, class A.
  • Marks and inscriptions in white enamel
  • Certificate of conformity with batch number
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
Informations de commande: Batch number and batch certificate included.