Imprimer
Acide perchlorique 65-71% (par analyse titrimétrique), TraceSELECT™ pour analyses de traces, Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
77227-100ML 77227-500ML 77227-1L
77227-100MLEA 162 EUR
77227-100ML 77227-500ML 77227-1L
Acide perchlorique 65-71% (par analyse titrimétrique), TraceSELECT™ pour analyses de traces, Fluka™
Acide perchlorique
TraceSELECT™ Ultra and TraceSELECT
  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
Formule: HClO₄
Point d'ébullition: 203 °C
Point de fusion: –18 °C
Densité: 1,66 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011325
Numéro CAS: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Assay 65 - 71 %
Silver (Ag) Max. 1 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 0.5 ppb
Barium (Ba) Max. 1 ppb
Beryllium (Be) Max. 1 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 10 ppb
Cadmium (Cd) Max. 1 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Indium (In) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 5 ppb
Manganese (Mn) Max. 1 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 20 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 0.5 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Tin (Sn) Max. 1 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 0.5 ppb
Zinc (Zn) Max. 5 ppb
Zirconium (Zr) Max. 2 ppb