Imprimer
Nalgene® Media Bottles, Square, Amber, with Screw Cap
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
322021-0125 322021-0030 322021-0060
NALG322021-0125EA 263 EUR
NALG322021-0125 NALG322021-0030 NALG322021-0060
Nalgene® Media Bottles, Square, Amber, with Screw Cap
Flacons Flacons pour milieux de culture
PETG, amber, translucent, with HDPE screw cap.
  • Non-sterile
  • Non-pyrogenic
  • Non-cytotoxic
Ideal for storage and shipping of light sensitive liquid media, buffers, and sera. Heavy-walled bottle provides an excellent gas barrier; features molded-in graduations, with a drip-proof neck finish and molded-in neck ring.
Heavy-walled, shatter resistant and durable, space-saving square design, molded in graduations, nonsterile, non-pyrogenic, non-cytotoxic, leakproof, with drip-proof neck finish and molded-in neck ring, PETG provides excellent gas barrier, with transparency and reduced permeability to CO₂ and O₂, amber bottle and closure provide protection for light sensitive reagents.
Certifications: Meets current standards for USP class VI, USP 661, and european pharmacopeia for abnormal toxicity. PETG media bottles and HDPE closure materials meet current biological testing standards including USP class VI.