Imprimer
Bouteilles rondes à col étroit, en FEP, avec bouchons à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
381600-0004 381600-0008 381600-0016 381600-0032
215-4082EA 773 EUR
215-4082 215-4083 215-4084 215-4085
Bouteilles rondes à col étroit, en FEP, avec bouchons à vis, Nalgene®
Flacons Flacons en plastique
Bottles, narrow neck, made of FEP, translucent, with ETFE screw cap.
  • Very good chemical resistance
  • Good temperature resistance
  • Linerless closure
Sample and store active pharmaceutical ingredients and bulk intermediates optimally with low particulate/low metals bottles with screw cap, certified to contain less than 20 particles per ml at 0,3 µm and greater. Metals certified to micrograms per liter (ppb) levels. Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.