Imprimer
Flacons, col large, carrés, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312114-0002 312114-0006 312114-0008 312114-0016 312114-0032
NALG312114-0002EA 1370 EUR
NALG312114-0002 NALG312114-0006 NALG312114-0008 NALG312114-0016 NALG312114-0032
Flacons, col large, carrés, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
HDPE, translucent, with PP screw cap.
  • Compatibility with most alcohols, bases and acids
  • Heat resistant up to +120 °C
  • Leakproof
Ideal for storing and shipping samples. Wide mouth permits easy filling and cleaning. Square shape saves space.
Easily fill and pour with Nalgene™ square wide-mouth HDPE bottles with closure. Featuring molded-in graduations for easy measuring and durable, uniform walls that are resistant to splitting or puncturing, these translucent bottles provide visible liquid levels and come in a space-saving square design.