Imprimer
Flacons, col large, conditionnement, avec bouchons à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312187-0001 312187-0002 312187-0004 312187-0008 312187-0016 312187-0032
NALG312187-0001EA 0 EUR
NALG312187-0001 NALG312187-0002 NALG312187-0004 NALG312187-0008 NALG312187-0016 NALG312187-0032
Flacons, col large, conditionnement, avec bouchons à vis, Nalgene®
Flacons Flacons en plastique
Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals with Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure. These durable, autoclavable bottles are designed for shipping and storage.
  • Very good chemical resistance
  • Very good temperature resistance
To ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.