Imprimer
Dropping bottles, LDPE, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312751-9025 312750-9050 312750-9125 312751-9125 312750-9025 312751-9050
215-2350EA 1010 EUR
215-2350 NALG312750-9050 NALG312750-9125 NALG312751-9125 NALG312750-9025 NALG312751-9050
Dropping bottles, LDPE, Nalgene®
Flacons Flacons compte-gouttes
Dropper bottles made up of natural/white low density polyethylene.
  • Round shape
  • Drop size: 40/50 μl
  • Leakproof
Certifications: Compliance with ISO 13485:2003; 21CFR177.1520 and USP Class VI.
Informations de commande: Controlled-drop dispensing tip and coloured closures are available seperately.