Hydrogène peroxyde 29 - 32% (p/p) en solution aqueuse stabilisé ACS
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
CAS No.: 7722-84-1
Molecular Formula: H2O2
Formula Weight: 34.02
Physical Form: Liquid
Density: 1.11
Refractive Index: 1.3350
Boiling Point: 107°
Solubility: Soluble in water, alcohol and ether. Insoluble in petroleum
Beilstein Reference: 3587191
Merck Reference: 14,4798
UN No.: 2014
MDL No.: MFCD00011333
For bleaching and deOdourizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagent
Formule:
H₂O₂ Point d'ébullition: 107 °C Point de fusion: < 0 °C Densité: 1,11 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00011333 Numéro CAS: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1,II Indice Merck: 14,4798 |