Imprimer
Bottles, economy, narrow neck, round, HDPE, with screw caps, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS2085-0004 DS2085-0032
331-4223EA 115 EUR
331-4223 331-4226
Bottles, economy, narrow neck, round, HDPE, with screw caps, Nalgene®
Bottles Plastic Bottles
HDPE, amber with amber PP screw cap.
  • Reduce UV light transmissions to protect light-sensitive liquids
  • Lightweight
  • Good chemical and temperature resistance
Conduct light-duty, general purpose applications where a heavier bottle is not required with these bottles.