Imprimer

1,2-Dichloroethylene-[D₂] (98% D)

Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): 1,2-Dichloroethylene-[²H₂]

CILDLM19360.1 CIL-DLM-1936-1
PROOCILDLM19360.1EA 748 EUR
PROOCILDLM19360.1 PROOCIL-DLM-1936-1
1,2-Dichloroethylene-[D₂] (98% D)
1,2-Dichloroethylene-[D₂]
Formule: C₂D₂Cl₂
Numéro CAS: 15075-90-8

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR