Imprimer
Nalgene® Packaging Bottles, Narrow Neck, Round, HDPE, Amber, with Screw Caps
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312084-0004 312084-0016
NALG312084-0004EA 645 EUR
NALG312084-0004 NALG312084-0016
Nalgene® Packaging Bottles, Narrow Neck, Round, HDPE, Amber, with Screw Caps
Flacons Flacons en plastique
HDPE, translucent amber, with PP screw cap.
  • Excellent impact and chemical resistance
  • For light sensitive materials
  • Leakproof
Robust construction; designed for storage, shipping, and packaging of light sensitive liquids.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Cdt: Bottles and closures are packed in separate polybags within the same carton.