Imprimer
Papier pour séparation de phases, grade 480
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FT-3-602-090 FT-3-602-185 FT-3-602-150 FT-3-602-125 FT-3-602-110 FT-4-602-185 FT-4-602-150 FT-4-602-125
SARTFT-3-602-090EA 25 EUR
SARTFT-3-602-090 SARTFT-3-602-185 SARTFT-3-602-150 SARTFT-3-602-125 SARTFT-3-602-110 SARTFT-4-602-185 SARTFT-4-602-150 SARTFT-4-602-125
Papier pour séparation de phases, grade 480
Filtres Papiers filtre Papiers séparateurs de phase
This phase separating paper is impregnated with stabilised silicon, thus rendering it hydrophobic and it retains water, but allows solvents to flow through. The flow stops automatically when the entire solvent has passed through, this eliminates the need to use separating funnels.
Used for filtration of organic solvents that have been contaminated with water to separate the aqueous from the organic phase, filtration of extracting solvents in clinical or medical labs and separation of emulsions that are formed during the extraction of aqueous plant or drug solutions.